Would you like to log in again?
DuPont™ Tychem® 10000 Encapsulated Level B Suits
Supplier: DuPont
Tychem®
TK527TLY3X000100 TK527TLYSM000100 TK527TLYMD000100 TK527TLY2X000100 TK527TLYXL000100 TK527TLY4X000100 TK527TLYLG000100
76187-680CS 573.08 USD InStock
76187-680 76187-890 76187-892 76187-674 76187-676 76187-678 76187-888
DuPont™ Tychem® 10000 Encapsulated Level B Suits
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Encapsulated Level B suit
  • Expanded back
  • Front entry
  • Standard visor
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust vents
  • Taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment. Coveralls also provide coverage from overhead liquid splash (does not provide vapor protection). Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Standard face shield is made of 40 mil PVC. Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Expanded back accommodates self-contained air breathing apparatus (SCBA). Liquid resistant, extra-long zipper increases garment opening and aids in donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Elastic opening at wrists allows for a tighter fit. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty