Would you like to log in again?
DuPont™ Tychem® 5000 Coveralls with Respirator Hood and Attached Gloves and Socks
Supplier: DuPont
Tychem®
C3185TTNMD000600 C3185TTNLG000600 C3185TTN2X000600 C3185TTN4X000600 C3185TTNXL000600 C3185TTNSM000600 C3185TTN3X000600
76187-354CS 1461.9 USD InStock
76187-354 76187-322 76187-356 76187-324 76187-326 76187-328 76187-736
DuPont™ Tychem® 5000 Coveralls with Respirator Hood and Attached Gloves and Socks
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Respirator hood
  • Attached dual-layer gloves
  • Multi-layer laminate interior gloves
  • Neoprene exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook & loop closure
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls are tan.
Tychem® CPF3 HD garments were designed with extensive input from first responders to ensure that HAZMAT, law enforcement, military and hospital personnel get the protection they need from chemical warfare agents (including Sarin, Mustard, and Lewisite) and toxic industrial chemicals (TIC's). Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator. Larger zipper pull is easier to grasp for easier donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five layer laminate film for a broad range of chemical protection. Neoprene outer gloves deliver high tactility and dexterity to handle tools or weapons. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty