Would you like to log in again?
DuPont™ Tychem® 10000 Encapsulated Training Suits with Serged Seams, Front Entry
Supplier: DuPont
Tychem®
TK586SLY3X000100 TK586SLYXL000100 TK586SLY4X000100 TK586SLY2X000100 TK586SLYLG000100 TK586SLYMD000100
76187-738CS 431.55 USD InStock
76187-738 76188-038 76187-740 76187-742 76187-744 76188-040
DuPont™ Tychem® 10000 Encapsulated Training Suits with Serged Seams, Front Entry
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Labeled as a training suit
  • Expanded back with front entry
  • Extra-wide visor
  • Single layer
  • Attached butyl gloves
  • Attached socks with outer boot loops
  • Storm flap
  • Serged seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These training suits are lime yellow.
Tychem® 10000 fully encapsulated training suits offer an economical and effective option for HazMat training. These suits are made from the same fabric and garment design as standard Tychem® 10000 suits. Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Extra wide (EX) single layer faceshield (20 mil PVC) allows for wide viewing angles. Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Integrated butyl gloves and internal adjustable waist belt system allow for support and improved fit.
Certifications: Thickness meets ASTM D1777. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: These suits are clearly labeled for training use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty