Would you like to log in again?
DuPont™ ProShield® 50 Coveralls with Open Wrists and Ankles
Supplier: DuPont
ProShield®
NB120SWH5X002500 NB120SWH3X002500 NB120SWH4X002500 NB120SWH2X002500 NB120SWHSM002500 NB120SWHMD002500 NB120SWH6X002500 NB120SWHLG002500 NB120SWHXL002500
76187-518CS 117.23 USD InStock
76187-518 76187-804 76187-806 76187-808 76187-520 76187-810 76187-812 76187-802 76187-516
DuPont™ ProShield® 50 Coveralls with Open Wrists and Ankles
Disposable Garments
ProShield® 50 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Collared
  • Open wrists and ankles
  • Serged seams
ProShield® 50 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate.
Applications include: janitorial, sanitation, and general industrial maintenance.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty