Would you like to log in again?
DuPont™ ProShield® 50 Coveralls with Standard Hood and Attached Skid Resistant Boots
Supplier: DuPont
Pro Shield
NB122SWHXL002500 NB122SWH2X002500 NB122SWH4X002500 NB122SWHSM002500 NB122SWHLG002500 NB122SWH6X002500 NB122SWHMD002500 NB122SWH5X002500 NB122SWH3X002500
76187-816CS 149.03 USD InStock
76187-816 76187-818 76187-522 76187-820 76187-524 76187-822 76187-526 76187-824 76187-814
DuPont™ ProShield® 50 Coveralls with Standard Hood and Attached Skid Resistant Boots
Disposable Garments
ProShield® 50 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Standard fit hood
  • Elastic wrists
  • Attached skid-resistant boots
  • Serged seams
ProShield® 50 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include: janitorial, sanitation, and general industrial maintenance.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty