Would you like to log in again?
DuPont™ Tychem® RESPONDER® CSM Encapsulated Level A Suits
Supplier: DuPont
RC550TTN2X00017C RC550TTNLG00 RC550TTN2X00017S RC550TTNXS00017W RC550TTNXL00017W RC550TTNLG00017W RC550TTNLG00017C RC550TTNMD00 RC550TTN4X00017W RC550TTNLG00017S RC550TTN5X00017C RC550TTNSM00017C RC550TTN4X00017S RC550TTNMD00017S RC550TTNXL000100 RC550TTN4X00017C RC550TTN4X000100 RC550TTNMD000100 RC550TTN5X000100 RC550TTN2X000100 RC550TTN3X00 RC550TTNLG000100 RC550TTNSM000100 RC550TTNXL00017S RC550TTN2X00017W RC550TTNXL00017C RC550TTN3X00017C RC550TTN3X00017S RC550TTN3X000100 RC550TTNMD00017W RC550TTNXL00 RC550TTNXS000100 RC550TTNSM00 RC550TTNMD00017C RC550TTN2X00 RC550TTN3X00017W
13452-716CS 3700.43 USD InStock
13452-716 76187-570 CA13452-708 13452-714 76187-572 76187-550 13452-712 76187-552 76187-574 76187-498 13452-710 76187-554 76187-576 76187-578 76187-556 76187-558 CA13452-710 CA13452-712 CA13452-714 13452-708 CA13452-716 76187-580 CA13452-718 76187-582 76187-560 76187-562 76187-564 76187-840 76187-500 76187-566 76187-568 76187-502 76187-546 76187-548 76187-504 13452-718
DuPont™ Tychem® RESPONDER® CSM Encapsulated Level A Suits
Disposable Garments
Tychem® RESPONDER® CSM combines multiple film barriers laminated to both sides of a high-strength polypropylene substrate for high level protection against toxic and corrosive chemicals.
  • Expanded back
  • Front entry
  • Standard visor
  • Attached butyl gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust valves
  • Double-taped seams
It is effective for handling chemical warfare agents as well as military site cleanup and HazMat first response. Tychem® RESPONDER® CSM fabric provides at least 30 minutes of protection against 320 chemical challenges. Each suit undergoes rigorous inspection and each Level A suit, pressure testing. Each fabric lot is tested using live toxic agents plus one garment for each test fabric production run is also tested for seam barrier with live toxic agents. Coveralls are a desert tan color for low visibility applications.
The encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing. Faceshields are triple-layered (PVC 40 mil/Teflon® 5 mil/PVC 20 mil) and standard width. Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Expanded back accommodates self-contained air breathing apparatus (SCBA). Coveralls feature a vapor tight zipper, which is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion. Two exhaust valves release higher pressure inside encapsulating garments, while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1117. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Passthroughs are optional. Coveralls available with either (p/t) MSA CAMDS #491335 connector on right side, (p/t) Scott® #803620-01 Hansen fitting on right side, or (p/t) Interspiro #33689006 on right front.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty