Would you like to log in again?
DuPont™ Tychem® 6000 Coveralls with Respirator Hood and Attached Socks and Gloves
Supplier: DuPont
Tychem®
TF186TGY5X0006PI TF186TGYSM0006PI TF186TGYMD0006PI TF186TGY2X0006PI TF186TGY3X0006PI TF186TGY4X0006PI TF186TGYXL0006PI TF186TGYLG0006PI
76187-870CS 1789.42 USD InStock
76187-870 76187-872 76187-862 76187-874 76187-864 76187-876 76187-866 76187-868
DuPont™ Tychem® 6000 Coveralls with Respirator Hood and Attached Socks and Gloves
Disposable Garments
Tychem® 6000 garments are made of a proprietary barrier film laminated to a Tyvek® substrate.
  • Respirator fit hood
  • Attached socks with outer boot flaps
  • Attached butyl gloves
  • Storm flap
  • Taped seams
The fabric provides at least 30 minutes of protection against over 181 challenge chemicals including chemical warfare agents and toxic industrial chemicals. Tychem® 6000 garments are strong, durable and lightweight, and they are available in both high-visibility orange and low-visibility gray fabrics, making them a preferred choice for law enforcement, emergency medical services (EMS) technicians and military personnel.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Three-piece hood design with longer zipper that extends to chin allows for complete coverage of neck area and good fit around a respirator. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Attached gloves are butyl. Integrated socks are composed of the same garment material.
Packaging: Coveralls are packaged individually, 6 per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty