Would you like to log in again?
DuPont™ Tychem® 10000 Encapsulated Level A Suits with Standard Visor
Supplier: DuPont
Tychem®
TK552TLYLG00017R TK552TLYXL00017R TK552TLY2X00017R
76187-894CS 2376.58 USD InStock
76187-894 76187-896 76187-898
DuPont™ Tychem® 10000 Encapsulated Level A Suits with Standard Visor
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Encapsulated Level A suit
  • Flat back
  • Front entry
  • Standard visor
  • Attached dual-layered gloves
  • Multi-layer laminate interior gloves
  • Butyl exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust vents
  • Double taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Standard width faceshield is triple-layered (PVC 40 mil/Teflon® 5 mil/PVC 20 mil). Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Flat back design is for use with airline. Vapor tight zipper is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five layer laminate film for a broad range of chemical protection. Butyl B 161 exterior gloves provide mechanical and physical durability to help protect inner glove. Gloves are attached with vapor tight ring and clamp system and are replaceable. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Internal adjustable waist belt system allows for support and improved fit. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1777. Positive air pressure integrity meets ASTM F1052. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Requires passthrough. Coveralls feature MSA dual purpose #495670 Hansen fitting on left front waist.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty