Would you like to log in again?
DuPont™ Tychem® 10000 Encapsulated Level A Suits with EX Extra Wide Visor
Supplier: DuPont
Tychem®
TK555TLYLG00015V TK555TLYXL00015V TK555TLY2X000100 TK555TLYMD00015C TK555TLY3X000100 TK555TLYMD00017S TK555TLYMD00015V TK555TLY4X00015V TK555TLY4X000100 TK555TLY4X000100 TK555TLYXL00015C TK555TLY3X00 TK555TLY3X00015V TK555TLY3X00015C TK555TLY2X000100 TK555TLYLG000100 TK555TLYXL00017S TK555TLYLG00017S TK555TLYXL000100 TK555TLYXL000100 TK555TLY4X00015C TK555TLY3X00017S TK555TLY4X00017S TK555TLY2X00017S TK555TLYMD000100 TK555TLY2X00015C TK555TLYLG00017M TK555TLY2X00017M TK555TLYLG000100 TK555TLYMD000100 TK555TLYLG00015C TK555TLY2X00015V TK555TLYXL00017M
10010-352CS 1906.87 USD InStock
CA10010-362 10010-352 76188-018 76187-990 10010-356 76188-016 76187-992 CA10010-360 76188-036 10010-354 76188-014 76188-012 76188-034 76188-032 10010-358 76188-030 76187-910 76187-912 76187-906 76187-908 10010-362 CA10010-352 10010-360 CA10010-354 76188-028 76188-026 76188-024 76187-986 76188-022 76188-020 CA10010-356 76187-988 CA10010-358
DuPont™ Tychem® 10000 Encapsulated Level A Suits with EX Extra Wide Visor
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Encapsulated Level A suit
  • Expanded back
  • Rear entry
  • Extra-wide EX visor
  • Attached dual-layered gloves
  • Multi-layer laminate interior gloves
  • Butyl exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust vents
  • Double taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing. Expanded faceshield provides wide 220 degree viewing angle. EX faceshield is triple-layered (laminate composed of 40 mil PVC/5 mil Teflon® /20 mil PVC). Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment. Expanded back to accommodate self-contained air breathing apparatus (SCBA). Coveralls also feature a vapor tight zipper that is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five layer laminate film for a broad range of chemical protection. Butyl B 161 gloves provide mechanical and physical durability to help protect inner glove. Gloves are attached with a vapor tight ring and clamp system and are replaceable. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Internal adjustable waist belt system provides additional support and improved fit. Additional barrier material is sewn in at knees and over taped for added abrasion resistance when kneeling or crawling. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1777. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Passthroughs are optional.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty