Would you like to log in again?
DuPont™ Tyvek® 400 Coveralls with Elastic Wrists and Ankles, Zipper Front Closure, Comfort Fit Design
Supplier: DuPont
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tyvek®
TY125SWH4X002500 TY125SWH5X002500 TY125SWH7X002500 TY125SWHXL0025VP TY125SWH2X002500 TY125SWH3X0006G1 TY125SWH2X0025VP TY125SWHLG0025VP TY125SWH3X0025VP TY125SWH4X0025VP TY125SWH4X0025NF TY125SWHMD0025NF TY125SWH2X002500 TY125SWHLG0006G1 TY125SWH6X002500 TY125SWHXL0025NF TY125SWH7X0025NF TY125SWHXL0006G1 TY125SWH5X002500 TY125SWHMD002500 TY125SWH3X002500 TY125SWHMD0006G1 TY125SWH6X0025NF TY125SWHMD00 TY125SWH5X0025VP TY125SWHXL002500 TY125SWH6X002500 TY125SWHXL002500 TY125SWH2X0025NF TY125SWH3X002500 TY125SWH5X0025NF TY125SWH4X0006G1 TY125SWH3X0025NF TY125SWH7X002500 TY125SWHLG002500 TY125SWHLG0025NF TY125SWH6X0025VP TY125SWHMD0025VP TY125SWHLG00 TY125SWH7X0025VP TY125SWH2X0006G1
76188-238CS 280.54 USD InStock
76188-238 47731-180 76188-236 76188-234 76188-232 76187-950 13452-874 76188-230 76187-952 13452-872 76187-930 76187-932 76187-954 76187-934 76187-956 76187-948 CA13452-872 CA13453-576 CA13453-574 CA13452-874 CA13453-572 47731-182 47731-184 76188-228 76188-226 76187-960 76187-940 76187-942 76188-240 76187-944 76187-946 76187-936 76187-958 76187-938 13453-578 13453-576 CA47731-184 13453-574 CA47731-180 13453-572 CA47731-182
DuPont™ Tyvek® 400 Coveralls with Elastic Wrists and Ankles, Zipper Front Closure, Comfort Fit Design
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Laydown collar
  • Zipper front
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Comfort fit design is based on extensive wearer input, to provide our most comfortable garment design that enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes longer zipper front for easier donning and doffing and an elastic waist to better position the garment. Coveralls feature a laydown collar, zipper front, and elastic wrists and ankles.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty