Would you like to log in again?
DuPont™ Tychem® 10000 Encapsulated Level A Suits with Extra Wide Visor and Fitting Options, Certified Compliant to NFPA 1994
Supplier: DuPont
Tychem®
TK612TLY2X00017N TK612TLY4X00017N TK612TLYLG00017W TK612TLY3X00017S TK612TLYMD00017S TK612TLY4X000100 TK612TLYMD00017W TK612TLY2X00017S TK612TLYXL00017N TK612TLYXL00017W TK612TLY5X000100 TK612TLY2X000100 TK612TLYXL000100 TK612TLYXS000100 TK612TLYLG00017N TK612TLYLG00017S TK612TLYXL00017S TK612TLYLG000100 TK612TLYMD00017N TK612TLY3X00017N TK612TLY4X00017W TK612TLY4X00017S TK612TLY2X00017W TK612TLY3X00017W TK612TLY3X000100 TK612TLYMD000100
76188-136CS 1867.46 USD InStock
76188-136 76188-078 76188-134 76188-076 76188-132 76188-074 76188-096 76188-130 76188-072 76188-094 76188-070 76188-092 76188-090 76188-128 76188-126 76188-124 76188-122 76188-088 76187-920 76188-086 76188-084 76187-914 76188-082 76187-916 76188-080 76187-918
DuPont™ Tychem® 10000 Encapsulated Level A Suits with Extra Wide Visor and Fitting Options, Certified Compliant to NFPA 1994
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Certified to NFPA 1994
  • Encapsulated Level A suit
  • Expanded back
  • Front entry
  • Extra-wide visor
  • Attached dual-layered gloves
  • Multi-layer laminate interior gloves
  • Neoprene exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust vents
  • Double taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Certified compliant to the 2012 edition of NFPA 1994, Class 2 Standard on Protective Ensembles for First Responders to CBRN Terrorism Incidents by an independent third-party certification organization. Class 2 garments are designed for situations where there is an immediate danger to life and health (IDLH) and must be worn with a self-contained breathing apparatus (SCBA). This is the highest level of protection under the NFPA 1994 standard.
Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Expanded face shield provides wide 220 degree viewing angle. Face shield is triple-layered (PVC 40 mil/Teflon® 5 mil/PVC 20 mil). Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Expanded back accommodates self-contained air breathing apparatus (SCBA). Vapor tight zipper is extra-long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five-layer laminate film for a broad range of chemical protection. Neoprene exterior gloves provide mechanical and physical durability to help protect inner glove. Gloves are attached with vapor tight ring and clamp system and are replaceable. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Internal adjustable waist belt system allows for support and improved fit. Additional barrier material is sewn at knees and over taped for added abrasion resistance when kneeling or crawling. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1777. Certified compliant to the 2012 edition of NFPA 1994. Positive air pressure integrity meets ASTM F1052. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Passthroughs are optional. Coveralls available with either (p/t) MSA Quick fill #80311 with cover on right side, (p/t) Scott® #803620-01 with Hansen fitting on right side, or (p/t) Interspiro #33689006 on right front
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty