Would you like to log in again?
DuPont™ Tychem® 10000 Encapsulated Level A Suits with Extra Wide Visor, Certified Compliant to NFPA 1994
Supplier: DuPont
Tychem®
TK613TLY4X000100 TK613TLYMD000100 TK613TLYXL000100 TK613TLYLG000100 TK613TLY2X000100 TK613TLY3X000100
76188-148CS 1867.46 USD InStock
76188-148 76188-138 76188-146 76188-144 76188-142 76188-140
DuPont™ Tychem® 10000 Encapsulated Level A Suits with Extra Wide Visor, Certified Compliant to NFPA 1994
Disposable Garments
Tychem® 10000 exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant.
  • Encapsulated Level A suit
  • Expanded back
  • Front entry
  • Extra-wide visor
  • Attached dual-layered gloves
  • Multi-layer laminate interior gloves
  • Neoprene exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook and loop closure
  • Two exhaust vents
  • Double taped seams
Tychem® 10000 fabric shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Tychem® 10000 is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. This high-performance chemical barrier fabric is suited for industrial, HazMat, and domestic preparedness applications. These coveralls are lime yellow.
Certified compliant to the 2012 edition of NFPA 1994, Class 2 Standard on Protective Ensembles for First Responders to CBRN Terrorism Incidents by an independent third party certification organization. Class 2 garments are designed for situations where there is an immediate danger to life and health (IDLH) and must be worn with a self-contained breathing apparatus (SCBA). This is the highest level of protection under the NFPA 1994 standard.
Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Expanded faceshield provides wide 220 degree viewing angle. Faceshield is triple-layered (PVC 40 mil/Teflon® 5 mil/PVC 20 mil). Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Expanded back accommodates self-contained air breathing apparatus (SCBA). Vapor tight zipper is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five layer laminate film for a broad range of chemical protection. Neoprene exterior gloves provide mechanical and physical durability to help protect inner glove. Gloves are attached with vapor tight ring and clamp system and are replaceable. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Internal adjustable waist belt system allows for support and improved fit. Additional barrier material is sewn at knees and over taped for added abrasion resistance when kneeling or crawling. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1777. Certified compliant to the 2012 edition of NFPA 1994. Positive air pressure integrity meets ASTM F1052. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Passthroughs are optional.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty