Would you like to log in again?
DuPont™ Tyvek® 400 Shirts
Supplier: DuPont
Tyvek®
TY303SWHMD005000 TY303SWH2X005000 TY303SWH2X0050VP TY303SWH5X005000 TY303SWH7X005000 TY303SWHXL0012G1 TY303SWH2X005000 TY303SWH4X005000 TY303SWHXL005000 TY303SWHLG0050VP TY303SWH4X0012G1 TY303SWH3X0050VP TY303SWH2X0012G1 TY303SWHMD005000 TY303SWHMD0012G1 TY303SWHXL005000 TY303SWHMD0050VP TY303SWH3X005000 TY303SWHSM005000 TY303SWHLG005000 TY303SWHLG0012G1 TY303SWH3X0012G1 TY303SWHLG005000 TY303SWHXL0050VP TY303SWH6X005000 TY303SWHSM005000 TY303SWH3X005000
10010-099CS 388.31 USD InStock
10010-099 10010-097 CA10010-101 76188-358 76188-356 76188-354 CA10010-107 76188-352 10016-936 CA10010-103 76188-350 10016-938 CA10010-105 10016-940 CA10010-099 76188-348 10010-103 76188-346 10010-101 76188-344 CA10010-097 76188-342 10016-942 10010-107 10010-105 76188-340 76188-360
DuPont™ Tyvek® 400 Shirts
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Laydown collar
  • Open wrists
  • Extends to hips
  • Front snap closure
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Shirt design features front snap closures for easy donning and doffing. Shirts also feature a laydown collar and open wrists. These shirts are typically worn with TY350S WH pants.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty