Would you like to log in again?
Proto® Torqueplus™ Metric Impact Sockets, 1", ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-10058M 577-10075M 577-10026M 577-10080M 577-10038M 577-10035M 577-10070M
76188-548EA 272.47 USD InStock
76188-548 76188-546 76188-536 76188-544 76188-534 76188-542 76188-528
Proto® Torqueplus™ Metric Impact Sockets, 1", ORS Nasco
Wrenches
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners.
  • Black oxide finish
  • Square drive
  • Pin locking type
  • Forged alloy steel
  • Metric measuring system
Proto Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty