Would you like to log in again?
Proto® Tether-Ready Offset Double Box Wrenches, Essendant
Supplier: Stanley
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-1077M-TT 577-1053M-TT 577-1172-TT 577-1050M-TT 577-1173-TT 577-1079M-TT 577-1171-TT 577-1175-TT 577-1057M-TT 577-1065M-TT 577-1072M-TT 577-1064M-TT 577-1170-TT 577-1061M-TT 577-1176-TT 577-1051M-TT
76188-558EA 60.77 USD InStock
76188-558 76188-566 76188-556 76188-564 76188-586 76188-554 76188-562 76188-552 76188-560 76188-572 76188-570 76190-396 76189-168 76189-174 76189-166 76189-170
Proto® Tether-Ready Offset Double Box Wrenches, Essendant
Wrenches
  • Full polish finish provides corrosion resistance and easy to clean surfaces
  • 12 Point box end allows for fine indexing, helping to improve speed and access
  • Offset box end allows access to recessed fasteners and keeps hands clear of obstructions
  • Box ends incorporate the TorquePlus™ system
  • Fits 4, 6 and 12 Point fasteners
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty