Would you like to log in again?
Proto® Crowfoot Wrenches, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-4996CF 577-4936CF 577-4909MCF 577-4710CF 577-4976CF
76189-398EA 42.3 USD InStock
76189-398 76189-046 76189-400 76188-994 76189-122
Proto® Crowfoot Wrenches, ORS Nasco
Wrenches
Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning.
  • Open opening type
  • Forged alloy steel
  • Inch measuring system
  • Ball locking type
  • Square drive
  • Nickel chrome finish
Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach. Full polish finish provides corrosion resistance and easy-to-clean surfaces.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty