Would you like to log in again?
Proto® Torqueplus™ Metric Combination Wrench Sets, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-1200RM-T50 577-1200HM-T50
76189-204BX 336.64 USD InStock
76189-204 76188-590
Proto® Torqueplus™ Metric Combination Wrench Sets, ORS Nasco
Wrenches
Increased torque applied to fastener with fewer rounded edges.
  • Forged alloy steel
  • Metric measuring system
  • Polish finish
  • Off-set handle
  • Includes tool roll
  • Open/box opening type
Unique arc design increases surface contact. Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty