Would you like to log in again?
Proto® Internal Pipe Wrenches, Essendant
Supplier: Stanley
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-140-38
76189-312EA 18.61 USD InStock
76189-312
Proto® Internal Pipe Wrenches, Essendant
Wrenches
Ideal for removing broken threaded pipe ends pipe plugs and fittings.
  • Installation and removal without thread damage
  • Eccentric cam action engages wrench against the walls of the pipe
  • Hex body allows for use of adjustable wrench for added torque
  • Made from hardened steel and plated to help prevent rust
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty