Would you like to log in again?
Proto® Angle Impact Wrenches, ORS Nasco
Supplier: ORS Nasco
577-150WA 577-138WA
76189-304EA 481.76 USD InStock
76189-304 76189-322
Proto® Angle Impact Wrenches, ORS Nasco
Wrenches
Low profile head improves access to tight areas while housing design contains gearing to reduce maintenance issues.
  • Trigger forward/reverse control
Impact mechanism provides high torque with little reaction, improving safety and reducing effort. Feather start trigger and forward/reverse ring for ease-of-use and increased productivity. Exhaust deflectors with integrated tethering connections for reduced noise and improved safety when working at heights. AJ2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty