Would you like to log in again?
Proto® 15 Piece Crowfoot Wrench Sets, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-49003CF 577-53001CF
76190-848EA 1664.1 USD InStock
76190-848 76189-392
Proto® 15 Piece Crowfoot Wrench Sets, ORS Nasco
Wrenches
Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning.
  • Open opening type
  • Forged alloy steel
  • Inch measuring system
  • Full polish
Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach. Full polish finish provides corrosion resistance and easy-to-clean surfaces. Locking hole feature helps keep the crowfoot wrench engaged on the extension during use, reducing downtime due to detachment of crowfoot in awkward spaces.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty