Would you like to log in again?
Proto® Torqueplus™ Sockets, ¹/₂", ORS Nasco
Supplier: ORS Nasco
577-5417 577-5408T 577-5410H 577-5408H
76189-524EA 15.12 USD InStock
76189-524 76189-550 76189-506 76189-508
Proto® Torqueplus™ Sockets, ¹/₂", ORS Nasco
Wrenches
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners.
  • Forged alloy steel
  • Square drive
  • Nickel chrome finish
  • Inch measuring system
  • Ball locking
Proto Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty