The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
- Manufactured with four detent holes; pin detent will engage correctly regardless of the direction the socket is placed on the square drive
- Positive Control System requires a separate tool to attach or remove the socket, meaning it cannot be removed without specific intent
- Chrome finish is applied after the holes are bored for better corrosion resistance
- Ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials
The TorquePlus™ configuration employs a radius-corner design, shifting contact area and stress away from the fastener corners and distributing contact stresses over a large area of the socket, minimizing slippage and fastener rounding, helping to extend fastener life, and allowing the user to more easily turn worn, rounded, or rusted fasteners.