Would you like to log in again?
Proto® Socket Hex Bits, ORS Nasco
Supplier: ORS Nasco
577-075903/4 577-49905/16 577-47703/16 577-54411/4 577-075905/8 577-54413/8 577-54415/16 577-49901/8 577-49901/4 577-54415/8 577-49907/32 577-47705/64 577-47705/32 577-47709/64 577-54417/16 577-47707/64 577-49905/32 577-47701/8 577-47701/16 577-075907/8 577-54413/4 577-49903/16 577-075909/16 577-47703/32 577-47701/4 577-47705/16 577-54419/16 577-49903/8 577-47707/32 577-54411/2
76188-514EA 64.06 USD InStock
76188-514 76189-638 76189-618 76189-644 76189-028 76189-646 76189-024 76189-048 76189-026 76189-104 76189-020 76189-022 76189-066 76189-084 76189-092 76188-522 76188-486 76189-628 76189-634 76189-018 76189-612 76189-630 76189-036 76189-038 76189-032 76189-056 76189-034 76189-030 76188-518 76189-074
Proto® Socket Hex Bits, ORS Nasco
Wrenches
Heat-treated.
  • Forged alloy steel
  • Square drive type
  • Inch measuring system
  • Hex tip type
Precision machined tip for proper screw engagement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty