Would you like to log in again?
Proto® Newton Meter/Inch Pound Dial Torque Wrenches, ORS Nasco
Supplier: ORS Nasco
577-6113NMF
76189-722EA 481.2 USD InStock
76189-722
Proto® Newton Meter/Inch Pound Dial Torque Wrenches, ORS Nasco
Wrenches
Accurate to ± 4% of torque reading in both directions at 20% to 100% of full scale.
  • Easy-to-read, high contrast scale
Memory needles are standard on all wrenches except electric light models. Torsion drive design--accuracy not effected by hand position. Dual scale (Metric/English) with clockwise and counterclockwise operation--fewer wrenches needed to complete job.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty