Would you like to log in again?
Proto® Foot Pound/Meter Kilogram Dial Torque Wrenches, Essendant
Supplier: Stanley
577-6134F 577-6149F
76189-724EA 1146.24 USD InStock
76189-724 76189-726
Proto® Foot Pound/Meter Kilogram Dial Torque Wrenches, Essendant
Wrenches
Memory needle pinpoints peak torque.
  • Dual scales
  • Ball locking
  • Male square drive
  • Alloy steel
  • Inch/metric
Easy to read dials. Withstands up to 20% accidental overload without losing calibration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty