Would you like to log in again?
Proto® Spline Reversing Ratcheting Wrenches, Essendant
Supplier: Stanley
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-SCV40 577-SCV22 577-SCVM19 577-SCV18 577-SCVM17 577-SCVM06 577-SCVM32 577-SCVM13 577-SCV10 577-SCV28 577-SCVM10 577-SCV48 577-SCVM34 577-SCV16 577-SCVM27 577-SCVM07 577-SCVM21 577-SCVM15 577-SCVM14 577-SCV07 577-SCVM12 577-SCV09 577-SCV14 577-SCV11 577-SCVM11 577-SCV20 577-SCVM22 577-SCV12 577-SCV24 577-SCVM09 577-SCVM30 577-SCVM16 577-SCV34 577-SCV30 577-SCV08 577-SCVM18 577-SCV26 577-SCV36 577-SCVM08 577-SCVM24 577-SCV32
76191-312EA 29.85 USD InStock
76191-312 76190-192 76190-170 76191-316 76190-290 76190-152 76190-174 76190-250 76190-272 76190-278 76190-330 76190-198 76190-254 76190-210 76190-232 76190-216 76190-312 76190-334 76190-158 76190-236 76190-318 76190-338 76191-310 76190-180 76190-240 76190-284 76190-186 76190-260 76190-140 76190-244 76190-266 76190-300 76190-146 76190-322 76190-220 76190-342 76190-204 76190-326 76190-224 76190-228 76190-306
Proto® Spline Reversing Ratcheting Wrenches, Essendant
Wrenches
The 15° angled offset box end helps keep shank clear of obstructions.
  • Black chrome finish
  • Open/box opening
Anti-Slip Design (ASD) open end. Black chrome finish is corrosion resistant and visually distinctive. Loads are kept off the corners of the fastener which helps prevent fastener rounding. Patented locking grooves help prevent the wrench from slipping off the fastener when turning. The arched surfaces are designed to spread the contact forces over a greater area. The chrome-vanadium switch lever allows for easy changes in turning direction which helps improve speed.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty