Would you like to log in again?
Proto® Torqueplus™ 6-Point Short Combination Wrenches, ORS Nasco
Supplier: ORS Nasco
577-1216EF 577-1204EF 577-1220EF
76188-626EA 27 USD InStock
76188-626 76188-596 76190-474
Proto® Torqueplus™ 6-Point Short Combination Wrenches, ORS Nasco
Wrenches
Torqueplus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Chrome finish
  • Oval handle
  • Open/box opening
  • Inch measuring system
  • Short length group
  • Forged alloy steel
Reduces risk of fastener rounding. Applies more torque to fastener. Unique arc design increases surface contact. Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury. Shank is positioned at the center of box end, reduces the tendency of the wrench to cock.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty