Would you like to log in again?
Proto® Tether-Ready Drive Deep Sockets, ORS Nasco
Supplier: ORS Nasco
577-5414-TT 577-5415M-TT 577-5014M-TT 577-5442-TT 577-5207M-TT 577-5326-TT 577-5322-TT 577-5420-TT 577-5424-TT 577-5208-TT 577-5023M-TT 577-5328-TT 577-5006M-TT 577-5316-TT 577-5423M-TT 577-5010M-TT 577-5020M-TT 577-5009M-TT 577-5444-TT 577-5216M-TT 577-5012M-TT 577-5214-TT 577-5008-TT 577-5330-TT 577-5216-TT 577-5212M-TT 577-5013M-TT 577-5420M-TT 577-5424M-TT 577-5413M-TT 577-5438-TT 577-5334-TT 577-5016M-TT 577-5412-TT 577-5440-TT 577-5314-TT 577-5022-TT 577-5024-TT 577-5218-TT 577-5422-TT 577-5318-TT 577-5320-TT 577-5421M-TT 577-5021M-TT 577-5015M-TT 577-5215M-TT 577-5230-TT 577-5220-TT 577-5324-TT 577-5416-TT 577-5008M-TT 577-5210M-TT 577-5218M-TT 577-5419M-TT 577-5410M-TT 577-5208M-TT 577-5212-TT 577-5417M-TT 577-5431M-TT 577-5418-TT 577-5206M-TT 577-5211M-TT 577-5336-TT 577-5427M-TT 577-5418M-TT 577-5430-TT 577-5342-TT 577-5217M-TT 577-5332-TT 577-5422M-TT 577-5007M-TT 577-5221M-TT 577-5338-TT 577-5226-TT 577-5429M-TT 577-5017M-TT 577-5232-TT 577-5210-TT 577-5436-TT 577-5222-TT 577-5026-TT 577-5219M-TT 577-5222M-TT 577-5228-TT 577-5411M-TT 577-5213M-TT 577-5011M-TT 577-5019M-TT 577-5414M-TT 577-5018M-TT 577-5416M-TT 577-5225M-TT 577-5412M-TT 577-5010-TT 577-5022M-TT 577-5340-TT 577-5312-TT 577-5428-TT 577-5209M-TT 577-5223M-TT 577-5426-TT 577-5214M-TT 577-5434-TT 577-5224M-TT 577-5432-TT 577-5220M-TT 577-5016-TT 577-5430M-TT 577-5226M-TT 577-5224-TT
76190-772EA 39.16 USD InStock
76190-772 76190-770 76190-776 76189-128 76190-774 76189-526 76189-486 76189-244 76189-246 76189-488 76190-778 76189-482 76189-240 76189-484 76189-480 76190-780 76190-782 76189-534 76189-414 76189-536 76190-786 76189-134 76189-530 76189-652 76189-136 76189-654 76189-258 76189-130 76189-494 76189-252 76189-132 76189-254 76189-490 76189-250 76189-492 76189-538 76189-418 76189-424 76189-546 76189-548 76189-420 76189-542 76189-422 76189-544 76189-148 76189-144 76189-540 76189-260 76189-428 76189-556 76189-436 76189-558 76189-438 76190-802 76189-156 76189-552 76189-158 76190-800 76189-554 76189-434 76189-152 76190-804 76189-154 76189-430 76190-850 76190-854 76189-602 76190-852 76189-442 76190-736 76189-564 76189-444 76190-734 76190-856 76189-560 76189-164 76189-440 76189-160 76189-608 76190-740 76189-606 76190-744 76189-574 76190-748 76189-576 76189-610 76189-570 76189-572 76190-790 76190-794 76190-792 76190-798 76190-796 76189-464 76189-586 76189-466 76189-588 76189-582 76189-580 76189-238 76189-476 76189-234 76189-598 76189-478 76189-236 76189-592 76189-472 76189-594 76189-590 76189-470
Proto® Tether-Ready Drive Deep Sockets, ORS Nasco
Tool Sets
The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
  • Manufactured with four detent holes; pin detent will engage correctly regardless of the direction the socket is placed on the square drive
  • Positive Control System requires a separate tool to attach or remove the socket, meaning it cannot be removed without specific intent
  • Chrome finish is applied after the holes are bored for better corrosion resistance
  • Ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials
The TorquePlus™ configuration employs a radius-corner design, shifting contact area and stress away from the fastener corners and distributing contact stresses over a large area of the socket, minimizing slippage and fastener rounding, helping to extend fastener life, and allowing the user to more easily turn worn, rounded, or rusted fasteners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty