Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® Hard Hat Lanyards, Pack of 10, Stanley® Products
Supplier: Stanley
Proto®
577-LANHATRD10
76191-098EA 123.44 USD InStock
76191-098
Proto® Hard Hat Lanyards, Pack of 10, Stanley® Products
Fall Protection Fall Lanyards and Ropes
Lanyards can be used with almost any hard hat, and they are a comfortable alternative to the traditional "chin strap''.
Great way to prevent a hard hat from becoming a dropped object. While keeping the helmet secure, the lanyard attaches to an outer garment, never impeding on the comfort or use of the hard hat. All Proto® hard hat lanyards feature an "easy release" to disengage the side release buckle, leaving one piece secured to the hard hat and the other still attached to an outer garment. When ready to use the hard hat again, simply re-engage the side release buckle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty