Would you like to log in again?
Proto® Super Heavy-Duty Offset Slugging Wrenches, Essendant
Supplier: Stanley
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-USN328 577-USN339 577-USN322 577-USN334 577-USN333 577-USN345
76191-336EA 90.76 USD InStock
76191-336 76191-338 76190-394 76190-392 76191-340 76191-342
Proto® Super Heavy-Duty Offset Slugging Wrenches, Essendant
Wrenches
Heavy box walls provide greater torque transmission
  • Black oxide finish
  • Box opening
  • Forged alloy steel
  • Inch measuring system
  • Off-set handle
High strength rectangular design withstands heavy sledging. Straight handle resists springing for easier nut turning. For close quarter work where hammer or sledge is required to loosen frozen nuts or set large fasteners.
Box opening remains on fastener when wrench is struck.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty