Would you like to log in again?
Left Handed Mayo Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-1555LFT
76191-388EA 149.7 USD InStock
76191-388
Left Handed Mayo Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar® also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty