Would you like to log in again?
SPME Fibers and Arrows
Supplier: Thermo Scientific
36SP01P1 36SP10P3 36SP01P3 36SP06E3 36SP03P1 36SP05T3 36SP03M5 36SP06E1 36SP09C1 36SP10P1 36SP03P3 36SP08A1 36SP05T1 36SP09C3
76191-410PK 1691.19 USD InStock
76191-410 76191-432 76191-422 76191-434 76191-412 76191-414 76191-404 76191-416 76191-406 76191-428 76191-418 76191-408 76191-430 76191-420
SPME Fibers and Arrows
Chromatography Supplies Chromatography Spare Parts
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step using Thermo Scientific™ SPME Arrow and SPME Fiber products. These products are used with the Thermo Scientific™ TriPlus RSH™ autosampler prior to gas chromatography (GC) or gas chromatography mass spectrometry (GC/MS) analysis.
The SPME Arrow and Fiber products are designed for volatile and semi-volatile compounds having a large range of boiling point and polarity. This sample preparation technique is ideal for ultra-trace and trace GC and GC/MS analysis of solid, liquid, or gas samples from environmental, food, clinical, and forensic industries.
The SPME Arrow and Fiber comprise a thin fused-silica fiber coated by an organic phase. They extract and concentrate analytes by ab- or adsorption and desorption processes. The fiber is successively thermally desorbed into a GC injector and the analytes are transferred to the analytical column. Selecting SPME Fiber or SPME Arrow depends on hardware compatibility and which product is used in your validated laboratory method.
SPME Fibers:
Original proven design used by many customers in standard methods
Wide selection of sorption materials
Requires SPME fiber conditioning module
SMART SPME fibers are available for automatic usage tracking if used with the TriPlus RSH SMART autosampler
SPME Arrow:
Improved, optimized fiber geometry
Faster extraction, higher throughput, more robust
Wide selection of sorption materials
Requires compatible firmware, SPME Arrow Conditioning Module and Heatex Stirrer, and an adaptation kit for the GC inlet of the TRACE1300 Series GC Instant Connect Split Splitless injector to accommodate the larger diameter of SPME Arrows
SMART SPME Arrow Fibers are available for automatic usage tracking if used with the TriPlus RSH SMART autosampler
SPME Arrow and SPME Fiber products are for use with the TriPlus RSH autosampler with GC analysis using the Thermo Scientific TRACE 1300 Series GC with an Instant Connect Split/Splitless injector or a TRACE GC Ultra Split/Splitless injector. Please select the Support button near the top of this page to ask Thermo Fisher Scientific representative about compatibility with other GC manufacturers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty