Would you like to log in again?
Caspar Screwdriver, Sklar
Supplier: Sklar
13-7052
76191-480EA 985.16 USD InStock
76191-480
Caspar Screwdriver, Sklar
Screwdrivers
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Caspar Screwdriver is for use with Caspar distraction screws to hold vertebral implant plates in place. It is a pentagonal shaped screwdriver that fits over part of the screw versus simply into the tip. It has an overall length of 8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty