Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fiber-optic Wisconsin Blades, Sklar®
Supplier: Sklar
07-1259 07-1260 07-1258
76191-554EA 400.47 USD InStock
76191-554 76191-764 76191-552
Fiber-optic Wisconsin Blades, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Wisconsin Laryngoscope Blades are most commonly used for tracheal intubation. The Wisconsin blade is straight with a slightly curved tip to facilitate placement of an endotracheal tube. It also has a wider, C-shaped flange, which makes it useful with infants and children with large tongues.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty