Would you like to log in again?
Gradle Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3444
76191-578EA 335.61 USD InStock
76191-578
Gradle Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Sklarhone™ Gradle Scissors, similar to Iris scissors, are used in ophthalmologic, dermatological and micro surgery. They feature one knife-like razor sharp edge and one standard scissor edge, allowing the user to make the sharp incisions of a knife with the cutting ability of scissors. The razor edge blades with sharp tips and are good for dissecting and cutting delicate tissue. The Sklarhone™ line of scissors are visuially identifiable by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty