Would you like to log in again?
Utility Sterilization Forceps, OR-Grade, Sklar®
Supplier: Sklar
10-1081
76191-624EA 73.56 USD InStock
76191-624
Utility Sterilization Forceps, OR-Grade, Sklar®
Forceps
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Sterilization Forceps are a versatile utility instrument most commonly used for placing and removing instruments within a sterilization container. The circular, paddle-style tips are serrated so they are particularly useful for grasping a variety of instruments. This product has a non-ratcheted, wireform finger ring handle for easy control. One finger ring is slightly larger than the other, allowing multiple fingers to fit for even greater control. The curved jaws are longer than the shanks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty