Would you like to log in again?
Mixter Scissors, OR Grade, Sklar
Supplier: Sklar
15-1361 15-1360
76191-666EA 205.18 USD InStock
76191-666 89025-496
Mixter Scissors, OR Grade, Sklar
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Mixter Scissors are medium weight scissors used for dissection of medium weight tissue. Available both straight and curved, the jaws of Mixter scissors are 1/2 the length of the shanks.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty