Would you like to log in again?
Blade Removal Forceps, OR-Grade, Sklar®
Supplier: Sklar
06-3091
76191-716EA 401.02 USD InStock
76191-716
Blade Removal Forceps, OR-Grade, Sklar®
Forceps
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Sklar's® Blade Removal Forceps are a non-ratcheted, finger ring instument used for safely removing scalpel blades from their handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty