Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fiber-optic Macintosh Laryngoscope Blade, English Style, Sklar®
Supplier: Sklar
07-1445 07-1444 07-1442 07-1441 07-1440 07-1443
76191-752EA 356.61 USD InStock
76191-752 76191-754 76191-756 76191-758 76191-760 76191-750
Fiber-optic Macintosh Laryngoscope Blade, English Style, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Macintosh Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. In general, Macintosh laryngoscope blades are curved. English Style Macintosh blades are longer than American Style Macintosh blades and feature a more continuous curvature. The English style blade tends to reveal a better view for the user, and is the preferred style to use if laryngoscopy is anticipated to be difficult.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty