Would you like to log in again?
Hi-Level Bandage Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3354
76191-762EA 353.01 USD InStock
76191-762
Hi-Level Bandage Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Hi-Level Bandage Scissors are non-ratcheted, finger ring scissors primarily used for removing tight bandages. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. They are identified by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty