Would you like to log in again?
Vidal Lupus Curette, OR-Grade, Sklar®
Supplier: Sklar
06-4090
76191-774EA 109.67 USD InStock
76191-774
Vidal Lupus Curette, OR-Grade, Sklar®
Curettes
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The Vidal Curette is commonly used in dermatological procedures to scrape and remove skin growths and lesions. It may also be known as a Lupus Curette. It features a smooth shovel-like tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty