Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fiber-optic Miller Laryngoscope Blade, Sklar®
Supplier: Sklar
07-1432 07-1430 07-1433 07-1431 07-1434 07-1435
76191-810EA 268.08 USD InStock
76191-810 76191-812 76191-814 76191-816 76191-818 76191-808
Fiber-optic Miller Laryngoscope Blade, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Miller Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. The Miller blade is straight and therefore longer than the curved Macintosh blade. It is inserted past the epiglottis, typically causing less trauma during intubation. This fiber-optic blade conforms to the Green System of fiber-optic compatibility which is indicated by a green bar at the posterior end of the blade where the blade attaches to a fiber-optic handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty