Would you like to log in again?
Lister Bandage Scissors, OR-Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15-3356 15-3355 15-3358
76191-804EA 272.47 USD InStock
76191-804 76191-826 76193-048
Lister Bandage Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Lister Bandage Scissors are a versatile instrument with multiple uses. They are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric, and occasionally umbilical cord. The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone™ instruments are identified by dual black finger rings.
Ring handles can be equal in size or with one ring larger. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone™ instruments are identified by dual black finger rings.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty