Would you like to log in again?
E-Z Lock Instrument Stringer, Sklar®
Supplier: Sklar
10-1826 10-1818 10-1834 10-1814
76193-098EA 239.15 USD InStock
76193-098 76191-830 76191-878 76191-662
E-Z Lock Instrument Stringer, Sklar®
Forceps and Instrument Holders
These are used to bundle instruments for sterilization.
  • Made of stainless steel
  • Industry standard
It is designed to keep an instrument in the open position in order to access all debris remaining after a procedure. The rectangular bar has one hinged end that opens to accommodate ringed instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty