Would you like to log in again?
Bergmann Bandage Scissors, OR-Grade, Sklar®
Supplier: Sklar
11-1378
76191-866EA 189.73 USD InStock
76191-866
Bergmann Bandage Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Bergmann Bandage Scissors are durable, lightweight, and designed for cutting thick dressing material. These bandage scissors have a guard on the lower blade and a blunt tip on the upper blade so as not to cause any injury to the patient while removing bandages or dressings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty