Would you like to log in again?
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Supplier: Sklar
10-1011
76191-892EA 166.23 USD InStock
76191-892
Cheatle Sterilization Forceps, OR-Grade, Sklar®
Forceps
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Cheatle Sterilization Forceps are a versatile utility instrument most commonly used for placing and removing instruments within a sterilization container. This product has a non-ratcheted finger ring handle for easy control of the instrument.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty