Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Standard Macintosh Blades, OR-Grade, Sklar®
Supplier: Sklar
07-1203 07-1201 07-1200 07-1202
76191-786EA 92 USD InStock
76191-786 76191-930 76191-778 76191-910
Standard Macintosh Blades, OR-Grade, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Macintosh Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. In general, Macintosh laryngoscope blades are curved. English Style Macintosh blades are longer than American Style Macintosh blades and feature a more continuous curvature. The English style blade tends to reveal a better view for the user, and is the preferred style to use if laryngoscopy is anticipated to be difficult. This blade is not fiber-optic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty