Would you like to log in again?
Silicone Instrument Holder for Cassettes, Sklar®
Supplier: Sklar
10-6535 10-6536 10-6538 10-6544 10-6533 10-6539 10-6543 10-6542 10-6540 10-6541 10-6531 10-6534 10-6537 10-6532 10-6545 10-6530
76191-960EA 19.83 USD InStock
76191-960 76191-972 76191-950 76191-962 76191-974 76191-952 76191-964 76191-976 76191-954 76191-966 76191-978 76191-956 76191-968 76191-958 76191-980 76191-970
Silicone Instrument Holder for Cassettes, Sklar®
Forceps and Instrument Holders
Silicone Instrument Holders are used for positioning instruments inside a cassette with a detachable lid used for cleaning, storage, or sterilization. All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The holders are repositionable to assist in storing different types and size instruments within a casette. They are available in a variety of colors and sold in sets of 3.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty