Would you like to log in again?
Sklar Edge™ Tungsten Carbide Lexer Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklar Edge™
16-2428 16-2436 16-2438 16-2426
76192-000EA 337.6 USD InStock
76192-000 76191-994 76191-996 76191-998
Sklar Edge™ Tungsten Carbide Lexer Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Lexer scissors can be used in multiple procedures and settings.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar Edge™ TC Lexer scissors have a standard beveled blade. Depending on physician preference, both the straight and the curved version may be used interchangeably, but the straight version is typically used for cutting surface tissue or sutures. Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. They can be identified by two gold coated finger rings.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty