Would you like to log in again?
Sklar Edge™ Tungsten Carbide Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
Sklar Edge™
16-2625 16-2610
89025-724EA 471.46 USD InStock
89025-724 76192-002
Sklar Edge™ Tungsten Carbide Dissecting Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Dissecting Scissors are used for dissecting and cutting tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar Edge™ Tungsten Carbide Scissors provide a durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors.
Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty